Check Order Status
Stainless Steel Lids For Test Sieve
Actual product may vary
Fisherbrand
Stainless Steel Lids For Test Sieve
Providing a secure closure to Fisherbrand™ Stainless Steel Test Sieves.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Outer
FKS11592573
100 mm
FKS11507033
200 mm
2 Options
Catalog number
FKS11592573
Price (SGD)
118.00
EA
Add to cart
Diameter (Metric) Outer:
100 mm
Price (SGD)
118.00
EA
Add to cart
Available in 100 and 200mm diameter sizes
Conforms to BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Specifications
Quantity
1 Ea.
Type
Sieve Cover
Material
Stainless Steel
For Use With (Application)
Providing closure to test sieves
Diameter (Metric) Outer
100 mm