Thermographic Measurements Temperature Strip
Thermographic Measurements Temperature Strip
Actual product may vary
Fisherbrand
Thermographic Measurements Temperature Strip
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS1159250377°C to 116°C
Catalog number FKS11592503
Price (SGD)
-
Temperature Range (Metric):
77°C to 116°C
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
For Use With (Application)For general purpose temperature monitoring
Product TypeTemperature Strip
Max. Temperature (Metric)116°C
Min. Temperature (Metric)77°C
Temperature Range (Metric)77°C to 116°C
Unit SizePK