Polypropylene Yellow Pipet Tip E
Polypropylene Yellow Pipet Tip E
Actual product may vary
Fisherbrand
Polypropylene Yellow Pipet Tip E
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Yellow Pipet Tips E are perfectly fit for manual or electronic, single or multichannel pipets.
Have Questions?
Catalog NumberVolume (Metric)
FKS115874421 to 200 μL
Catalog number FKS11587442
Price (SGD)
33.00
PK
Add to cart
Volume (Metric):
1 to 200 μL
Price (SGD)
33.00
PK
Add to cart
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Volume (Metric)1 to 200 μL
ColorYellow