Class B Amber Glass Threaded Vials with Attached Caps
Class B Amber Glass Threaded Vials with Attached Caps
Actual product may vary
Fisherbrand
Class B Amber Glass Threaded Vials with Attached Caps
Type I, Class B amber borosilicate glass
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Height (Metric)Diameter (Metric) OuterVolume (English)
FKS115835424 mL48 mm15 mm1 dr.
FIS14-955-3212 mL38 mm12 mm0.5 dr.
FKS115735422 mL38 mm12 mm0.5 dr.
FIS14-955-32240 mL98 mm28 mm10.82 dr.
FIS14-955-3314 mL48 mm15 mm1 dr.
FIS14-955-3328 mL63 mm17 mm2.16 dr.
FIS14-955-33320 mL60 mm28 mm5.4 dr.
FKS1150355220 mL60 mm28 mm5.4 dr.
Catalog number FKS11583542
Price (SGD)
258.00
EA
Add to cart
Volume (Metric):
4 mL
Height (Metric):
48 mm
Diameter (Metric) Outer:
15 mm
Volume (English):
1 dr.
Price (SGD)
258.00
EA
Add to cart
Type I, Class B amber borosilicate glass
  • General-purpose vials for light-sensitive materials
  • Made from Type I, Class B Amber Borosilicate Glass
  • Autoclavable
Specifications
ClassificationB
Material (Liner)Teflon-faced Rubber
ColorAmber
Diameter (Metric) Outer15 mm
MaterialAmber Glass (Class B)
SterilityNon-sterile
DisposableSingle-use
Volume (Metric)4 mL
For Use With (Application)For general purpose vials for light sensitive materials
No. per Pack144
Volume (English)1 dr.
Volume (Metric) Total4 mL
Closure MaterialPhenolic
AutoclavableAutoclavable
Closure TypeFitted Cap
Height (Metric)48 mm
Quantity144 Pk.
Certifications/ComplianceASTM, USP Type I
TypeVial
Closure Size13-425
Unit SizeEA