Type 1 Class A Borosilicate Glass Clear Sample Vial with Fitted Cap and Polycone Liner
Type 1 Class A Borosilicate Glass Clear Sample Vial with Fitted Cap and Polycone Liner
Actual product may vary
Fisherbrand
Type 1 Class A Borosilicate Glass Clear Sample Vial with Fitted Cap and Polycone Liner
Use these vials for general lab applications. Fisherbrand™ Type 1 Class A Borosilicate Glass Clear Sample Vial with Fitted Cap and Polycone Liner features low profile for greater stability and compound storage.
Have Questions?
Catalog NumberColor
FKS11583522Clear
Catalog number FKS11583522
Price (SGD)
202.00
EA
Add to cart
Color:
Clear
Price (SGD)
202.00
EA
Add to cart
  • Supplied in partitioned trays
  • Standard vials have fitted black phenolic caps with either polycone liner or PTFE faced white rubber liner
  • Also available: pre-cleaned EPA (Environmental Protection Agency) vials for VOA (volatile organic analysis), with or without certification, having fitted white polypropylene open top caps with PTFE faced silicone septa
Specifications
ClassificationA
Material (Liner)Polycone
ColorClear
Diameter (Metric) Outer28 mm
MaterialBorosilicate Glass (Type 1 Class A)
Volume (Metric)20 mL
No. per Pack72
Closure MaterialPhenolic
Closure TypeFitted Cap
Height (Metric)60 mm
Quantity72 Pk.
Closure Size24-400
Unit SizeEA