Glass Conical Separating Funnel with PTFE Stopcock
Glass Conical Separating Funnel with PTFE Stopcock
Actual product may vary
Glass Conical Separating Funnel with PTFE Stopcock
Manufactured from Borosilicate Glass and ISO 4800 Certified. Fisherbrand™ Glass Conical Separating Funnel with PTFE Stopcock is supplied with a glass and plastic stopper. Available with interchangeable glass or PTFE stopcock.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Stopper Number
FKS11582782250 mL24/29
FKS11572782100 mL19/26
Catalog number FKS11582782
Price (SGD)
172.00
EA
Add to cart
Capacity (Metric):
250 mL
Stopper Number:
24/29
Price (SGD)
172.00
EA
Add to cart
Manufactured from Borosilicate Glass and ISO 4800 Certified. Fisherbrand™ Glass Conical Separating Funnel with PTFE Stopcock is supplied with a glass and plastic stopper. Available with interchangeable glass or PTFE stopcock.
  • Manufactured from Borosilicate Glass
  • ISO 4800 Certified
  • Supplied with a glass and plastic stopper
  • Available with interchangeable glass or PTFE stopcock
Specifications
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Certifications/ComplianceISO 4800
Quantity1PK
StopcockPTFE
Stopper MaterialGlass, Plastic
Stopper Number24/29
Product TypeSeparatory Funnel
Unit SizeEA