Temperature Strip
Temperature Strip
Actual product may vary
Fisher Catalogue
Temperature Strip
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS1158250340°C to 71°C
Catalog number FKS11582503
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
40°C to 71°C
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
Max. Temperature (Metric)71°C
Min. Temperature (Metric)40°C
Temperature Range (Metric)40°C to 71°C
Accuracy0.02
Subdivision5°C
Unit SizePK