Made from stainless steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has an 100 mm diameter.
Have Questions?
Change view
Catalog Number
Pore Size
FKS11582153
400 μm
FKS11532143
75 μm
FKS11512163
600 μm
FKS11562143
100 μm
FKS11542153
250 μm
5 Options
Catalog number FKS11582153
Price (SGD)
157.00
EA
Add to cart
Pore Size:
400 μm
Price (SGD)
157.00
EA
Add to cart
100 mm diameter test sieves
Stainless steel frame
Woven wire mesh
Supplied with Certificate of Conformity and record card
Specifications
Quantity1 Ea.
Pore Size400 μm
TypeSieve
Certifications/ComplianceCertificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000