Check Order Status
40mm PP Screw Seal, Black, Closed top, Assembled septum
Actual product may vary
Fisherbrand
40mm PP Screw Seal, Black, Closed top, Assembled septum
Polypropylene caps are chemically inert and suitable for most chromatography applications.
Have Questions?
Catalog Number
Color
FKS11580605
Black
Catalog number
FKS11580605
Price (SGD)
198.00
EA
Add to cart
Color:
Black
Price (SGD)
198.00
EA
Add to cart
40mm
PP
Screw Cap
Closed top
Combination Seal
Pre-assembled caps and septa are convenient and minimize contamination from handling
Specifications
Material (Liner)
PTFE Virginal
Color
Black
Diameter (Metric) Outer
40 mm
Material
Polypropylene
For Use With (Application)
Storage
Quantity
100
Thickness (Metric)
0.5 mm
Type
Screw Cap