Microtiter Tubes and Racks
Microtiter Tubes and Racks
Actual product may vary
Fisherbrand
Microtiter Tubes and Racks
Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeSterilityFormat
FKS11579944Microtiter TubeSterileRobotic Rack
3487Microtiter TubeSterileRobotic Rack
FIS21-403-208Microtiter TubeSterileRobotic Rack
Catalog number FKS11579944
Price (SGD)
214.00
PK
Add to cart
Product Type:
Microtiter Tube
Sterility:
Sterile
Format:
Robotic Rack
Price (SGD)
214.00
PK
Add to cart
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight-plug strips can be easily cut apart for individual samples
  • 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
  • Rigorous testing to ensure sample integrity and security
Packaging Options
  • Microtiter tubes and caps are available sterile and nonsterile
Specifications
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
ColorNatural
Volume (Metric)1.2 mL
Quantity96 tubes/rack 10 racks/unit, 5 units case
Closure MaterialPolyethylene (Plug)
Product LineThermo Scientific™
SterilitySterile
FormatRobotic Rack
Product TypeMicrotiter Tube
Unit SizePK