Check Order Status
Small Hub Removable Needle, Custom Length
Actual product may vary
Small Hub Removable Needle, Custom Length
Suitable for use with removable (RN)-style syringes thar are 100μL or smaller. Hamilton™ Small Hub Removable Needle, Custom Length is available in lengths between 0.375 and 12 inches.
Have Questions?
Catalog Number
Needle Gauge
FKS11573314
27 G
Catalog number
FKS11573314
Price (SGD)
2,077.00
PK
Add to cart
Needle Gauge:
27 G
Price (SGD)
2,077.00
PK
Add to cart
Autoclavable
Gas sterilizable
6/Pk
Specifications
Needle Gauge
27 G
Length (English)
0.375 to 12 in.
Product Type
Small Hub Needle
Needle Point Style
Custom Point Style (2, 3, or 4)