Solid Rubber Stoppers
Solid Rubber Stoppers
Actual product may vary
Fisherbrand
Solid Rubber Stoppers
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) BottomStopper NumberHeight (Metric)Diameter (Metric) Top
FKS1157292221 mm2128mm24.5 mm
FKS1154292215 mm1524mm18 mm
FKS1151293229 mm2932mm33 mm
Catalog number FKS11572922
Price (SGD)
54.00
PK
Add to cart
Diameter (Metric) Bottom:
21 mm
Stopper Number:
21
Height (Metric):
28mm
Diameter (Metric) Top:
24.5 mm
Price (SGD)
54.00
PK
Add to cart
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
  • Temperature range -40° to +70°C
  • Not suitable for use with petroleum oils and hydrocarbon solvents
Specifications
For Use With (Application)Laboratory use
Height (Metric)28mm
Quantity20/Pk.
ColorRed
Diameter (Metric) Bottom21 mm
Diameter (Metric) Top24.5 mm
MaterialRubber
Product TypeSolid Stopper
Stopper Number21
Unit SizePK