Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits
Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits
Actual product may vary
Fisherbrand
Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)
FKS1157094010 mm0.79 mL/cm vol./length
FIS08-700-11315 mm1.8 mL/cm vol./length
Catalog number FKS11570940
Price (SGD)
-
Diameter (Metric):
10 mm
Length (Metric):
0.79 mL/cm vol./length
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
Product TypeDialysis Trial Kit
MWCO0.1 to 0.5 kDa
MaterialCellulose Ester
Diameter (Metric)10 mm
Diameter (English)0.39 in.
Length (Metric)0.79 mL/cm vol./length
Kit Contents1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks
QuantityEach
For Use With (Application)Ideal for small, critical dialysis applications
Unit SizeEA
Contents & Storage
Recommended storage in refrigeration (4-8°C).