Soda Lime Glass Pasteur Pipette
Soda Lime Glass Pasteur Pipette
Actual product may vary
Fisherbrand
Soda Lime Glass Pasteur Pipette
Soda Lime Glass Pasteur Pipette is intended for single use in microbiological and serological laboratories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)TypeTip Style
FKS11566963230 mmUnplugged Pasteur PipetStandard Tip
FKS11546963150 mmUnplugged Pasteur PipetStandard Tip
FKS11506973150 mmCotton-plugged Pasteur PipetStandard Tip
Catalog number FKS11566963
Price (SGD)
149.00
CS
Add to cart
Length (Metric):
230 mm
Type:
Unplugged Pasteur Pipet
Tip Style:
Standard Tip
Price (SGD)
149.00
CS
Add to cart
  • Pasteur pipettes made of soda-lime glass are a good alternative to plastic pipettes
  • Complies with ISO 7712
  • Pasteur pipettes with short or long extended, fine tip (150 mm, 230 mm and 270 mm)
  • Pasteur pipettes with and without cotton plug
  • The filling volume is approx. 2 mL
  • Ideal for use with Fisherbrand Pipettors
Specifications
AutoclavableNot Autoclavable
MaterialSoda Lime Glass
For Use With (Application)Single Use in Microbiological and Serological Laboratory Applications, Pipetting Liquids, Dropwise Dosing of Liquids and Reagents, Distribution of Heated Liquid Quantities
Quantity1000 Pk.
Tip StyleStandard Tip
Volume (English)0.07 oz.
Volume (Metric)2 mL
Length (Metric)230 mm
TypeUnplugged Pasteur Pipet
DisposableSingle-use
For Use With (Equipment)Fisherbrand Pipetters
Unit SizeCS