Stainless Steel Cylindrical Scoops
Stainless Steel Cylindrical Scoops
Actual product may vary
Fisherbrand
Stainless Steel Cylindrical Scoops
Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Cylindrical Scoops are an ideal accessory to have if you own a weighing scale.
Have Questions?
Catalog NumberdimCapacityMetricLength (Metric)
FKS11563452260 mm
Catalog number FKS11563452
Price (SGD)
104.00
EA
Add to cart
Length (Metric):
260 mm
Price (SGD)
104.00
EA
Add to cart
Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Cylindrical Scoops are an ideal accessory to have if you own a weighing scale.
  • Durable, stainless steel, highly polished, and easy to clean cylindrical scoop 
  • Can be sterilised by autoclaving
  • Well-suited for pharmaceutical and environmental sampling
  • Ideal for weighing and handling of samples and other materials
  • Great accessory to have if you own a weighing scale
  • Perfect for weighing anything from liquid to gems and other materials
Specifications
Product TypeScoop
MaterialStainless Steel
IncludesWith Handle
StyleCylindrical
Dimensions (L x W)260 x 75 mm
Length (Metric)260 mm
Width (Metric) Bowl75 mm
Unit SizeEA