Check Order Status
Silicon Platinum-cured Tubing
Actual product may vary
Fisherbrand
Silicon Platinum-cured Tubing
Ideal to use with peristaltic pump due to its higher tear strength and less vulnerability towards splitting or mechanical damage.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Outer
FKS11562563
5 mm
FKS11502573
9 mm
2 Options
Catalog number
FKS11562563
Price (SGD)
50.00
EA
Add to cart
Diameter (Metric) Outer:
5 mm
Price (SGD)
50.00
EA
Add to cart
Tube has no smell, taint or reaction with chlorine
Platinum cured tubing has higher clarity with no yellowing, as experienced with peroxide cured materials
60°IRHD ±5°C
FDA, BGA and WRC approved
Temperature range: -60°C to 200°C
Anti-adherent, hydrophobic
High electrical and ozone resistance
Less bacteriological build up
Can be used for food, laboratory, pharmaceutical and medical applications
Specifications
Certifications/Compliance
FDA, BGA and WRC approved
Material
Platinum-cured Silicone
Diameter (Metric) Outer
5 mm
Thickness (Metric)
1 mm
For Use With (Application)
For food, laboratory, pharmaceutical and medical applications
Color
Translucent