Spectra/Por™ Magnetic Weighted Closures
Spectra/Por™ Magnetic Weighted Closures
Actual product may vary
Fisherbrand
Spectra/Por™ Magnetic Weighted Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Have Questions?
Catalog NumberDiameter (Metric)
FKS1156090035 mm
Catalog number FKS11560900
Price (SGD)
486.00
PK
Add to cart
Diameter (Metric):
35 mm
Price (SGD)
486.00
PK
Add to cart
Spectra/Por Magnetic Closure Type:
  • Made from polypropylene with a fluorocarbon coated magnetic SS bar
  • Use on magnetic stir-plate to provide sample stirring for increased dialysis efficiency
  • 4 sealing width sizes: 23mm, 35mm, 55mm and 75mm
  • Autoclavable
  • 10/pkg
  • Use with RC dialysis tubing only
  • Not for use with CE tubing
Specifications
Product TypeDialysis Tubing Closure
MembraneRegenerated Cellulose
MaterialPolypropylene
Diameter (Metric)35 mm
AutoclavableAutoclavable
Closure TypeMagnetic
ColorRed
Temperature Range (English)32°F to 250°F
Temperature Range (Metric)0°C to 121°C
Kit ContentsMagnetic fluorocarbon coated stainless steel bar
Quantity2 Pack
For Use With (Application)Use on magnetic stir-plate to provide sample stirring for increased dialysis efficiency, use with RC dialysis tubing only, not for use with CE tubing
Unit SizePK