Class A Clear Glass Threaded Vials with Caps, PTFE-faced White 14B Rubber Liner
Class A Clear Glass Threaded Vials with Caps, PTFE-faced White 14B Rubber Liner
Actual product may vary
Fisherbrand
Class A Clear Glass Threaded Vials with Caps, PTFE-faced White 14B Rubber Liner
With PTFE-faced, white, 14B rubber liner
Have Questions?
Catalog NumberVolume (Metric)Height (Metric)Diameter (Metric) OuterClosure Size
FKS115535424 mL38 mm17 mm15-425
Catalog number FKS11553542
Price (SGD)
342.00
EA
Add to cart
Volume (Metric):
4 mL
Height (Metric):
38 mm
Diameter (Metric) Outer:
17 mm
Closure Size:
15-425
Price (SGD)
342.00
EA
Add to cart
With PTFE-faced, white, 14B rubber liner
Caps are attached.
  • General-purpose vials
  • Made from Type I, Class A borosilicate glass
  • Short style vials feature low profile for greater stability and compound storage
  • Packaged in partitioned trays
Specifications
ClassificationA
Material (Liner)Rubber
ColorClear
Diameter (Metric) Outer17 mm
MaterialBorosilicate Glass
SterilityNon-sterile
DisposableSingle-use
Volume (Metric)4 mL
For Use With (Application)For general purpose vials for light sensitive materials
Volume (English)1 dr.
Volume (Metric) Total4 mL
Closure MaterialPhenolic
Closure TypeScrew Cap
Height (Metric)38 mm
Certifications/ComplianceASTM, USP Type I
TypeVial
Closure Size15-425
Unit SizeEA