Check Order Status
Solid Natural Rubber Stoppers
Actual product may vary
Fisherbrand
Solid Natural Rubber Stoppers
Used for laboratory purposes.
Have Questions?
Catalog Number
Diameter (Metric) Bottom
Diameter (Metric) Top
FKS11552922
17 mm
20.3 mm
Catalog number
FKS11552922
Price (SGD)
42.00
PK
Add to cart
Diameter (Metric) Bottom:
17 mm
Diameter (Metric) Top:
20.3 mm
Price (SGD)
42.00
PK
Add to cart
Made up of natural rubber
Red in color
Solid stoppers with no holes
Specifications
For Use With (Application)
For laboratory use
Hardness
46/55 IRHD
Length (Metric)
26 mm
Quantity
20/Pk.
Color
Red
Diameter (Metric) Bottom
17 mm
Diameter (Metric) Top
20.3 mm
Material
Natural Rubber
Product Type
Solid Stopper