Check Order Status
Bossheads
Actual product may vary
Fisherbrand
Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Bossheads come with offset jaws for greater strength.
Have Questions?
Change view
Catalog Number
Includes
FKS11547722
Clamp Screw, Plastic Knobs
FKS11537722
Clamp Screw, Metal Knobs
2 Options
Catalog number
FKS11547722
Price (SGD)
154.00
PK
Add to cart
Includes:
Clamp Screw, Plastic Knobs
Price (SGD)
154.00
PK
Add to cart
Aluminum alloy with clamping screws
For use with rods up to 16mm in diameter
Specifications
Quantity
5/Pk.
Type
Bosshead Clamp
Material
Aluminum Alloy
Includes
Clamp Screw, Plastic Knobs
For Use With (Equipment)
For use with rod up to 16mm diameter