Class A Clear Glass Threaded Vials with Caps, PTFE-faced White 14B Rubber Liner
Class A Clear Glass Threaded Vials with Caps, PTFE-faced White 14B Rubber Liner
Actual product may vary
Fisherbrand
Class A Clear Glass Threaded Vials with Caps, PTFE-faced White 14B Rubber Liner
Fisherbrand™ Type 1 Class A Borosilicate Glass Clear Sample Vials with Fitted Caps feature fitted black phenolic caps with either polycone liner or PTFE faced white rubber liner.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Height (Metric)Diameter (Metric) Outer
FKS115435328 mL63 mm17 mm
FIS14-955-3162 mL38 mm12 mm
FIS14-955-32030 mL94 mm25 mm
FKS1159353230 mL94 mm25 mm
FKS115435422 mL31 mm15 mm
FIS14-955-3288 mL63 mm17 mm
FIS14-955-3256 mL43 mm19 mm
FKS115635426 mL43 mm19 mm
FIS14-955-3274 mL48 mm15 mm
FKS1151354220 mL60 mm28 mm
FIS14-955-32912 mL68 mm19 mm
Catalog number FKS11543532
Price (SGD)
285.00
EA
Add to cart
Volume (Metric):
8 mL
Height (Metric):
63 mm
Diameter (Metric) Outer:
17 mm
Price (SGD)
285.00
EA
Add to cart
Fisherbrand™ Type 1 Class A Borosilicate Glass Clear Sample Vials with Fitted Caps feature fitted black phenolic caps with either polycone liner or PTFE faced white rubber liner.
  • Supplied in partitioned trays
  • Also available: pre-cleaned EPA (Environmental Protection Agency) vials for VOA (volatile organic analysis), with or without certification, and fitted white polypropylene open top caps with PTFE faced silicone septa
Specifications
ClassificationA
Material (Liner)Teflon-faced Rubber
ColorClear
Diameter (Metric) Outer17 mm
MaterialBorosilicate Glass (Type 1 Class A)
SterilityNon-sterile
DisposableSingle-use
Volume (Metric)8 mL
For Use With (Application)For general purpose vials for light sensitive materials
No. per Pack144
Volume (English)2.16 dr.
Volume (Metric) Total8 mL
Closure MaterialPhenolic
Closure TypeFitted Cap
Height (Metric)63 mm
Quantity144 Pk.
Certifications/ComplianceASTM, USP Type I
TypeVial
Closure Size15-425
For Use With (Equipment)Volatile Organic Analysis
Unit SizeEA