Polystyrene Diamond-Shaped Antistatic Weighing Boat
Polystyrene Diamond-Shaped Antistatic Weighing Boat
Actual product may vary
Fisherbrand
Polystyrene Diamond-Shaped Antistatic Weighing Boat
Designed for accurate pouring with minimal sample loss. Fisherbrand™ Polystyrene Diamond-Shaped Antistatic Weighing Boat features a smooth surface that flexes easily. Disposable boats have a flat bottom, static-free surface and can be used for a variety of lab applications.
Have Questions?
Catalog NumberCapacity (Metric)ColorShape
FKS11543432100 mLWhiteDiamond
Catalog number FKS11543432
Price (SGD)
191.00
EA
Add to cart
Capacity (Metric):
100 mL
Color:
White
Shape:
Diamond
Price (SGD)
191.00
EA
Add to cart
  • Features a diamond shape that easily flexes for accurate pouring
  • Wide, flat bottom helps resist tipping
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
  • Temperature range -10 to 70°C
  • Antistatic surface to ensure complete sample removal
  • 500 FDA/food-grade compliant boats per pack
Specifications
Bottom ShapeFlat
Capacity (Metric)100 mL
ColorWhite
DisposableSingle-use
MaterialPolystyrene
ShapeDiamond
TypeAntistatic Weighing Boat
For Use With (Equipment)Microwave Moisture Analyzer
Dimensions (L x W x H)119 x 90 x 19 mm
Length (Metric)119 mm
Width (Metric)90 mm
Height (Metric)19 mm
Unit SizeEA