Rectangular Retort Stand Bases
Rectangular Retort Stand Bases
Actual product may vary
Fisherbrand
Rectangular Retort Stand Bases
Excellent for help holding onto bench equipment and glassware. Fisherbrand™ Rectangular Retort Stand Bases are manufactured from blue acrylic finished steel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS11538102250 mm
FKS11518102160 mm
FKS11528102200 mm
Catalog number FKS11538102
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
  • Without rod
  • With 10mm hole
  • Fitted with rubber feet
  • Threaded at the center of one short side
Specifications
MaterialStainless Steel
Dimensions (L x W)250 x 160 mm
Length (Metric)250 mm
IncludesWith Rubber Feet
Product TypeRetort stand base
ColorBlue
Unit SizeEA