Check Order Status
Universal pH Indicator Paper
Actual product may vary
Fisherbrand
Universal pH Indicator Paper
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
Have Questions?
Catalog Number
pH Range
FKS11537392
6.4 to 8
Catalog number
FKS11537392
Price (SGD)
59.00
PK
Add to cart
pH Range:
6.4 to 8
Price (SGD)
59.00
PK
Add to cart
Filter paper soaked with one or several colored indicators of pH
Available in a variety of formats to meet individual needs
Specifications
pH Range
6.4 to 8
Format
Roll
Quantity
4 Rolls
Type
Grade 911
Length (Metric)
5 m
Width (Metric)
7 mm