Stainless Steel Offset Spoons
Stainless Steel Offset Spoons
Actual product may vary
Fisher Catalogue
Stainless Steel Offset Spoons
The unique curved shape spoon, offset, allows contact with all corners of bottles and containers reducing sample losses. The alternate end can be used as a scraper to chip away at hard samples or used as a spatula for sampling or lifting.
Have Questions?
Catalog NumberLength (Metric)
FKS11533492190 mm
Catalog number FKS11533492
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
190 mm
  • Laboratory Offset Spoon
  • Stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Uses the finest in corrosion-resistant 304 stainless steel
  • The unique curved shape spoon, offset, allows contact with all corners of bottles and containers reducing sample losses 
  • The alternate end can be used as a scraper to chip away at hard samples or used as a spatula for sampling or lifting
  • Can be used within laboratory to retrieve powders, granules, solids and viscous liquids
Specifications
Quantity1 Ea.
SterilitySterile
AutoclavableAutoclavable
Product TypeLaboratory Offset Spoon
FinishBright
For Use With (Application)For Sterile Applications
Temperature (Metric) Max-196°C to +550°C
MaterialStainless Steel
Length (Metric)190 mm
Unit SizeEA