Check Order Status
Stainless Steel Dual Spoon/Spatulas
Actual product may vary
Fisherbrand
Stainless Steel Dual Spoon/Spatulas
Manufactured from heavyweight stainless steel. Fisherbrand™ Stainless Steel Dual Spoon/Spatula combines the effectiveness of two tools into one.
Have Questions?
Catalog Number
Length (Metric)
FKS11533462
100 mm
Catalog number
FKS11533462
Price (SGD)
96.00
PK
Add to cart
Length (Metric):
100 mm
Price (SGD)
96.00
PK
Add to cart
Manufactured from stainless steel
Dual use spatula/spoon combo
Specifications
Quantity
5 Pk.
Product Type
Spoon Spatula
Finish
Bright
Width (Metric)
10 mm
Diameter (Metric) Outer
3.6 mm
Dimensions (L x W)
100 x 10 mm
Material
Stainless Steel
Length (Metric)
100 mm