pH Electrode Storage Solution, Reagecon™
pH Electrode Storage Solution, Reagecon™
Actual product may vary
Fisherbrand
pH Electrode Storage Solution, Reagecon™
Designed to maintain proper functioning and accurate readings of pH and reference electrodes. pH Electrode Storage Solution, Reagecon™ is a 3M KCl solution that helps maintain proper working conditions after cleaning or when the electrode has not been in use for an extended time period.
Have Questions?
Catalog NumberQuantity
FKS11533182500 mL
Catalog number FKS11533182
Price (SGD)
-
Quantity:
500 mL
The response, accuracy and lifespan of pH electrodes can be substantially improved through correct handling, cleaning and storage. Designed for routine equipment maintenance, these solutions are safe to use with any sensitive electrode. Electrode storage solutions are made from potassium chloride, free from silver ion, and are suitable for all Blueline electrodes.
Specifications
CompositionPotassium Chloride solution, without Silver ion
Concentration3 mol/L KCl
For Use With (Equipment)pH and Reference Electrodes, All Blueline Electrodes
Filling Solution TypeElectrode Storage
Quantity500 mL
Unit SizeEA