Digital Colorimeter
Digital Colorimeter
Actual product may vary
Fisherbrand
Digital Colorimeter
Measure the absorbance of particular wavelengths of light by a specific solution Fisherbrand™ Digital Colorimeter with 240V transformer and filters.
Have Questions?
Catalog NumberRange
FKS115280920 to 199% (transmission)
Catalog number FKS11528092
Price (SGD)
4,178.00
EA
Add to cart
Range:
0 to 199% (transmission)
Price (SGD)
4,178.00
EA
Add to cart
  • Digital Colorimeter
  • With 240V transformer and filters
  • Filters 400 -700nm 
Specifications
Accuracy±0.05A at 1A
Spectral Bandwidth40 nm
TypeDigital Colorimeter
Voltage240 V
Wavelength Range400 to 700 nm
Electrical Requirements240 V, 50 Hz
Height (Metric)60 mm
Hertz50 Hz
Photometric Repeatability±0.02 A at 1 A
Range0 to 199% (transmission)
Resolution0.02 at 1 A (Absorbance), 2% (Transmission)
Width (Metric)146 mm
Unit SizeEA