Check Order Status
Silicone Tubing
Actual product may vary
Fisherbrand
Silicone Tubing
For consistently reliable and accurate fluid transfers.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
Thickness (Metric)
FKS11527083
8 mm
2 mm
FKS10664372
10.0 mm
2.0 mm
FKS11507083
6 mm
1.5 mm
3 Options
Catalog number
FKS11527083
Price (SGD)
199.00
EA
Add to cart
Diameter (Metric) Inner:
8 mm
Thickness (Metric):
2 mm
Price (SGD)
199.00
EA
Add to cart
Available in different diameters
Specifications
Diameter (Metric) Inner
8 mm
Overall Length (Metric)
10 m
Thickness (Metric)
2 mm