PTFE Magnetic Stir Bar, Cross-shaped
PTFE Magnetic Stir Bar, Cross-shaped
Actual product may vary
Fisherbrand
PTFE Magnetic Stir Bar, Cross-shaped
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS1152198310 x 10 x 5mm
Catalog number FKS11521983
Price (SGD)
-
Dimensions (L x W x H):
10 x 10 x 5mm
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
  • Stable, general-purpose magnetic stirrer
  • Provides a deep, quiet vortex even at relatively high speeds
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Dimensions (L x W x H)10 x 10 x 5mm
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Height (Metric)5mm
Length (English)0.375 in.
Width (English)0.375 in.
Width (Metric)10mm
CoatingPTFE
Length (Metric)10 mm
Quantity5/Pk.
ShapeCross
TypePTFE Stirrer Bars Cross
Unit SizePK