Sephadex™ LH-20
Sephadex™ LH-20
Actual product may vary
Fisherbrand
Sephadex™ LH-20
Use for molecular sizing of natural product such as steroid, terpenoid, lipid, low molecular weight peptide. Cytiva Life Sciences™Sephadex™ LH-20 is a liquid chromatography media useful to analytical and industrial scale for preparation of closely related molecular species.
Have Questions?
Catalog NumberQuantity
FKS11520784100 g
Catalog number FKS11520784
Price (SGD)
-
Quantity:
100 g
  • Prepared for use in polar organic solvents and aqueous solvent mixtures
  • Made from hydroxypropylated dextran beads that have been crosslinked to yield a polysaccharide network
  • Hydrophilic and lipophilic dual nature of the matrix enables powerful chromatographic selectivity
  • Batch-to-batch reproducibility
  • Partical size dry: 18μ-111μm
  • Fractionation range globular proteins: 4x103 to 5 x 103
  • Stable in most aqueous and organic eluent systems
  • Shelf life: 5years
  • Autoclavable: 20 min at 121°C
  • Flow Velocity <∽12cm/min
  • pH stability working range: 2-13
Analytical and industrial scales for the preparation of closely related molecular species
Specifications
Quantity100 g
Flow Rate720 cm/hr.
Particle Size18 to 111 μm in dry form
Product TypeSephadex LH-20 Media
FormatSolid Spherical Bead
Diameter (Metric) Inner2.6cm
For Use With (Application)For fractionation of lipids, hormones, vitamins, and other small biomolecules in polar organic solvents and aqueous solvent mixtures
Height (English)11.81 in.
Height (Metric)30cm
Max. Pressure25-45 cm/h, pressure drop cm H2O/bed height=2, beh height 30 cm, 2.6 cm i.d.
pH Range2 to 13
Support TypeHydroxypropylated, cross-linked Dextran
Sample TypeSteroid, Terpenoid, Lipid, low molecular weight Peptide
Unit SizeEA
Contents & Storage
4 to 30°C