Protein LoBind™ Deepwell Plates
Protein LoBind™ Deepwell Plates
Actual product may vary
Protein LoBind™ Deepwell Plates
Protein LoBind™ Deepwell Plates
Protein LoBind™ Deepwell Plates
Protein LoBind™ Deepwell Plates
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
Have Questions?
Catalog NumberColorNo. of WellsVolume (Metric) WellPackaging
FKS11518643White962000 μL5 Bags x 4 Plates
Catalog number FKS11518643
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
White
No. of Wells:
96
Volume (Metric) Well:
2000 μL
Packaging:
5 Bags x 4 Plates
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.
Specifications
FormatStandard
Array8 x 12
AutoclavableAutoclavable
Packaging5 Bags x 4 Plates
ColorWhite
Product TypeProtein LoBind™ Plate
MaterialPolypropylene
Volume (Metric) Well2000 μL
No. of Wells96
Volume (Metric) Working50 to 2,000 μL
SterilityNon-sterile
Dnase FreeYes
For Use With (Application)Proteomics or other fields of protein research
Max. RCF6000 x g
No. per Pack20 Pk.
Quantity20 Pk.
RNase FreeYes
Well ColorColorless
Dimensions (L x W x H)85.5 x 127.8 x 60 mm
Unit SizePK