Check Order Status
Two-Way Scaffolding Connectors
Actual product may vary
Fisherbrand
Two-Way Scaffolding Connectors
Used for fixing two rods or tubes at right angles. Fisherbrand™ Two-Way Scaffolding Connectors can be tightened by hand.
Have Questions?
Catalog number
FKS11518112
Price (SGD)
21.00
EA
Add to cart
Price (SGD)
21.00
EA
Add to cart
With thumbscrews
For rods and tubes of 6 to 13mm diameters
Specifications
Product Type
2-Way Connector
For Use With (Application)
2-Way