Check Order Status
Silicone Stoppers
Actual product may vary
Fisherbrand
Silicone Stoppers
For laboratory and/or food testing, industrial masking or educational use.
Have Questions?
Catalog Number
Diameter (Metric) Bottom
Quantity
FKS11517672
31 mm
2/Pk.
Catalog number
FKS11517672
Price (SGD)
71.00
PK
Add to cart
Diameter (Metric) Bottom:
31 mm
Quantity:
2/Pk.
Price (SGD)
71.00
PK
Add to cart
Non-toxic, translucent
Temperature range -60°C to 200°C
High electrical resistance
High resistance to many chemicals including ozone
Specifications
Quantity
2/Pk.
Diameter (Metric) Bottom
31 mm
Material
Silicone
Product Type
Stopper