Plain Economy PTFE Stir Bar
Plain Economy PTFE Stir Bar
Actual product may vary
Fisherbrand
Plain Economy PTFE Stir Bar
Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
FKS1151758250 mm8 mm
FKS1159757225 mm8 mm
FKS1150758240 mm8 mm
Catalog number FKS11517582
Price (SGD)
63.00
PK
Add to cart
Length (Metric):
50 mm
Diameter (Metric):
8 mm
Price (SGD)
63.00
PK
Add to cart
Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
CoatingPTFE
Diameter (Metric)8 mm
Length (Metric)50 mm
Quantity10/Pk.
TypePTFE Stirrer Bars Plain Economy
Unit SizePK