Stainless Steel Nuffield Spatulas
Stainless Steel Nuffield Spatulas
Actual product may vary
Fisherbrand
Stainless Steel Nuffield Spatulas
Manufactured from stainless steel. Fisherbrand™ Stainless Steel Nuffield Spatula is for sterile applications.
Have Questions?
Catalog NumberLength (Metric)
FKS11513492140 mm
Catalog number FKS11513492
Price (SGD)
14.28
Special offer
Ends: 15-Dec-2025
40.00
Save 25.72 (64%)
PK
Add to cart
Length (Metric):
140 mm
Price (SGD)
14.28
Special offer
Ends: 15-Dec-2025
40.00
Save 25.72 (64%)
PK
Add to cart
  • Nuffield pattern
  • Sterile
  • Stainless steel
  • Round, bent end and scoop end
  • Nuffield Spatula, 140mm, stainless steel
  • Supplied in standard packs of 10
Specifications
Quantity10 Pk.
SterilitySterile
AutoclavableAutoclavable
ShapeRound Bent End, Scoop End
Product TypeSpatula
For Use With (Application)For Sterile Applications
Temperature (Metric) Max-196°C to +550°C
MaterialStainless Steel
Length (Metric)140 mm
Unit SizePK