Nickel Chattaway Micro Spatulas
Nickel Chattaway Micro Spatulas
Actual product may vary
Fisher Catalogue
Nickel Chattaway Micro Spatulas
Get optimum results when you use the nickel-based micro spatulas for your essential laboratory work. Ideal for Biology, Microbiology and Pharmaceutical Applications, the Fisherbrand™ Nickel Chattaway Micro Spatulas are widely used, sturdy, affordable, and resistant to most solvents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS11513482200 mm
FKS11503482150 mm
Catalog number FKS11513482
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
200 mm
  • Excellent resistance to many corrosive environments
  • High thermal and electrical conductivities
  • Low gas content and low vapor pressure
  • Nickel 200 spec (minimum 99.6% nickel content)
  • Bright finish
  • Spatula Blade One end flat, other end at 45° angle
  • Supplied in standard packs of 5
Specifications
Quantity5 Pk.
Capacity (Metric)100 mL
Product TypeChattaway Spatula
Width (Metric)4 mm
Diameter (Metric) Outer3.6 mm
Dimensions (L x W)200 x 4 mm
MaterialNickel
Length (Metric)200 mm
Unit SizePK