Metal Case Uncontrolled Electromantle
Metal Case Uncontrolled Electromantle
Actual product may vary
Fisherbrand
Metal Case Uncontrolled Electromantle
The basic Thermo Scientific metal case uncontrolled mantle provides an element temperature of 450°C
Have Questions?
Catalog number FKS11512714
Price (SGD)
2,321.00
EA
Add to cart
Price (SGD)
2,321.00
EA
Add to cart
  • Basic (CMU) is available in twelve sizes from 50mL to 22 liters (22,000mL). The basic Thermo Scientific metal case uncontrolled mantle provides an element temperature 450°C (842°F)
  • All mantle types: Flexible coiled heating elements absorb shock, minimizing the risk of flasks breaking
  • Heating element is attached to a rigid thermal insulating cartridge to ensure maximum heat transfer and support
  • Heating elements and insulation form an easy-to-replace heating cartridge
  • Unique air flow through ventilation slots beneath and around the rim and heating cartridge keeps the exterior “cool-to-the-touch”
  • Stainless steel screen covering the heating element is grounded directly to the cable for added protection
  • Mantles are double fused for added safety
  • Non-skid feet and support clamps add stability
Specifications
Certifications/ComplianceCE
Capacity33.8 oz., 1000mL
Capacity (Metric)1000 mL
Height (English)6.69 in.
Voltage230 V
Wattage380w
Width (English)8.5 in.
Width (Metric)21.8cm
Product TypeHeating Mantle
Temperature (Metric)Ambient to 450°C
Amperage3.3 A
Capacity (English)33.8 oz.
Unit SizeEA