PCR Plate, 96-well, low profile, non-skirted
PCR Plate, 96-well, low profile, non-skirted
Actual product may vary
PCR Plate, 96-well, low profile, non-skirted
PCR Plate, 96-well, low profile, non-skirted
PCR Plate, 96-well, low profile, non-skirted
PCR Plate, 96-well, low profile, non-skirted
PCR Plate, 96-well, low profile, non-skirted
Fisherbrand
PCR Plate, 96-well, low profile, non-skirted
Low profile 96-well plates for use in PCR and qPCR applications.
Have Questions?
Catalog NumberQuantity
FKS1151029425 plates
Catalog number FKS11510294
Price (SGD)
-
Quantity:
25 plates
The low profile PCR plate has shorter wells than the standard profile plate (AB-0600), decreasing the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. These low profile products are especially recommended for use with reaction volumes below 20μL. They are available in assorted colors and are compatible with standard multichannel pipettes.
The low profile Thermo Scientific PCR plate has shorter wells than the standard profile plate (AB-0600), decreasing the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. These low profile products are especially recommended for use with reaction volumes below 20 μL. They are available in assorted colors and are compatible with standard multichannel pipettes.
Highlights
• Low profile to reduce dead space and to increase PCR efficiency
• Available with black alpha-numeric lettering
• Improved access for liquid handling
• Maximum fill volume of 0.2 mL
• Cut corner: H12
Applications
• PCR
• qPCR/Real-Time PCR
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified RNase/DNase-free
Packaging25 Plate
ColorClear
Product TypePCR Microplate
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Skirt StyleNon-skirt
MaterialPolypropylene
Volume (Metric) Well0.2 mL
No. of Wells96
Volume (Metric) Working0.1 mL
Quantity25 plates
Reaction SpeedLow profile
Unit SizePK