Octahedral PTFE Stir Bar
Octahedral PTFE Stir Bar
Actual product may vary
Fisherbrand
Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
FKS1150778238 mm8 mm
Catalog number FKS11507782
Price (SGD)
66.00
PK
Add to cart
Length (Metric):
38 mm
Diameter (Metric):
8 mm
Price (SGD)
66.00
PK
Add to cart
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action
  • All have pivot rings
  • AlNiCo-V magnets encased in PTFE
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
CoatingPTFE
Diameter (Metric)8 mm
Length (Metric)38 mm
Quantity10/Pk.
ShapeOctahedral
TypePTFE Stirrer Bars Octahedral
Unit SizePK