Conductivity Standard Solution, 1.3μS/cm, Reagecon™
Product Image
Fisherbrand
Conductivity Standard Solution, 1.3μS/cm, Reagecon™
Reagecon's Conductivity Standard at 25°C, is tested and certified by an ISO17025 Accredited Test Method (INAB Ref. 264T) and NIST traceable.
Have Questions?
Catalog NumberQuantity
FKS115067236 x 500 mL
Catalog number FKS11506723
Price (SGD)
3,015.00
PK
Add to cart
Quantity:
6 x 500 mL
Request bulk or custom format
Price (SGD)
3,015.00
PK
Add to cart
Specifications
DescriptionConductivity Standard Solution
PackagingBottles
Certificate of AnalysisYes
Conductivity1.3μS/cm ± 1%
Physical FormLiquid
View more
  • Certified by Reagecon's ISO 17025 Accredited Test Method (INAB Ref: 264T)
  • NIST traceable
  • This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the product label. The low temperature coefficient of variation, reduces measurement errors and enables non- temperature controlled use and is suitable for field work
  • High accuracy ± 1% at 25°C, enables the standards to be used as calibrators and/or controls, in accordance with the most exacting industrial requirements
  • The product is aqueous based, non-hazardous and accurate to a specification of ± 1%. The product because it is aqueous based, has a low temperature co-efficient of variation.