Conductivity solution HI-7034 c/w certificate of analysis 80,000μS/cm (μmho/cm) 460mL Hanna
Actual product may vary
Fisherbrand
Conductivity solution HI-7034 c/w certificate of analysis 80,000μS/cm (μmho/cm) 460mL Hanna
Have Questions?
Catalog Number
Capacity (Metric)
FKS11504337
460 mL
Catalog number FKS11504337
Price (SGD)
185.00
EA
Add to cart
Capacity (Metric):
460 mL
Price (SGD)
185.00
EA
Add to cart
A choice of ready mixed calibration standards traceable to NIST. Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air. Supplied in boxes of 25.