1x1 image pixel for data collection
Polystyrene Square-Shaped Standard Weighing Boat
Polystyrene Square-Shaped Standard Weighing Boat
Actual product may vary
Fisherbrand
Polystyrene Square-Shaped Standard Weighing Boat
Weigh and pour samples with ease. Fisherbrand™ Polystyrene Square-Shaped Standard Weighing Boat folds easily and features a smooth surface and round corners for easy pouring. Choose the size and color that works best for your needs. Disposable and durable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)TypeColor
FKS1150343285 mLAntistatic Weighing DishWhite
FKS1134412510 mLWeighing BoatWhite
FKS11364125280 mLWeighing BoatWhite
FKS1135412585 mLWeighing BoatWhite
Catalog number FKS11503432
Price (SGD)
102.00
PK
Add to cart
Capacity (Metric):
85 mL
Type:
Antistatic Weighing Dish
Color:
White
Price (SGD)
102.00
PK
Add to cart
Weigh and pour samples with ease. Fisherbrand™ Polystyrene Square-Shaped Standard Weighing Boat folds easily and features a smooth surface and round corners for easy pouring. Choose the size and color that works best for your needs. Disposable and durable.
  • Flexible boats feature smooth surfaces and rounded corners to provide accurate pour-outs with minimal sample loss
  • Select white, black or blue for best color contrast to ensure sample is completely removed
  • Boats are shallow with a wide, flat bottom to resist tipping
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases
  • Temperature range -10 to 70°C (14 to 158°F)
  • Available in three sizes and two colors, in packs of 500
  • All are FDA/food-grade compliant
Specifications
Bottom ShapeFlat
Capacity (Metric)85 mL
ColorWhite
DisposableSingle-use
MaterialPolystyrene
ShapeSquare
TypeAntistatic Weighing Dish
For Use With (Equipment)Microwave Moisture Analyzer
SizeMedium; 85 x 85 x 24 mm
Length (Metric)85 mm
Width (Metric)85 mm
Height (Metric)24 mm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link