Silicon Platinum-cured Tubing
Actual product may vary
Fisherbrand
Silicon Platinum-cured Tubing
Ideal to use with peristaltic pump due to its higher tear strength and less vulnerability towards splitting or mechanical damage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) Outer
FKS115025739 mm
FKS115625635 mm
Catalog number FKS11502573
Price (SGD)
151.00
Each
Diameter (Metric) Outer:
9 mm
Price (SGD)
151.00
Each
  • Tube has no smell, taint or reaction with chlorine
  • Platinum cured tubing has higher clarity with no yellowing, as experienced with peroxide cured materials
  • 60°IRHD ±5°C
  • FDA, BGA and WRC approved
  • Temperature range: -60°C to 200°C
  • Anti-adherent, hydrophobic
  • High electrical and ozone resistance
  • Less bacteriological build up
  • Can be used for food, laboratory, pharmaceutical and medical applications
Specifications
Certifications/ComplianceFDA, BGA and WRC approved
MaterialPlatinum-cured Silicone
Diameter (Metric) Outer9 mm
Thickness (Metric)1.5 mm
For Use With (Application)For food, laboratory, pharmaceutical and medical applications
ColorTranslucent
Unit SizeEach