Check Order Status
White Polyurethane Foam Stoppers
Actual product may vary
Fisherbrand
White Polyurethane Foam Stoppers
Used as convenient alternative method for closing sterile media flasks and vials instead of cotton plugs.
Have Questions?
Catalog Number
Diameter (Metric)
Size
FKS11502563
25mm
25 x 50mm
Catalog number
FKS11502563
Price (SGD)
36.00
PK
Add to cart
Diameter (Metric):
25mm
Size:
25 x 50mm
Price (SGD)
36.00
PK
Add to cart
Used for microbiology applications
During storage of media, it is preferable to cover the stoppers with aluminum foil to avoid dust contamination on removal
Shrinkage will occur after continued use following autoclaving, but this is dependent purely on application
To eliminate the possibility of obstructing the flow of air into the media container, care should be taken not to crease the stoppers on fitting
Specifications
Diameter (Metric)
25mm
For Use With (Application)
Microbiology
Length (Metric)
50 mm
Quantity
100/Pk.
Size
25 x 50mm
Color
White
Material
Polyurethane
Product Type
Stopper