Clifton™ Thermostatic Thermostirrer Water Baths
Clifton™ Thermostatic Thermostirrer Water Baths
Actual product may vary
Fisher Catalogue
Clifton™ Thermostatic Thermostirrer Water Baths
Obtain optimum immersion results for flasks, bottles, racks and other objects in a stable temperature environment. A Stainless steel tank contained within a sturdy outer case, this Clifton™ Thermostatic Thermostirrer Water Bath is robust and provides good water circulation and temperature stability.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1146950922 L
Catalog number FKS11469509
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
22 L
  • Stirred Clifton water baths are designed for immersion of flasks/bottles/racks in a stable temperature environment
  • Stainless steel tank contained within a sturdy outer case and stainless steel shelf provide good water circulation and temperature stability
  • The Thermostirrer is robust constructed from stainless steel for durability
  • Thermostirrer controls and painted surfaces feature “anti-bacterial technology
  • Painted surfaces feature a tough anti-bacterial finish with good chemical resistance
  • Easy to use controls wipe clean and have an anti-bacterial finish
  • Immersed components are corrosion resistant
Specifications
DescriptionDigital Water Bath
Capacity (Metric)22 L
Certifications/ComplianceCE Approved
Temperature Range5°C to 99°C
MaterialStainless Steel
ModelNE4-22 T
Temperature Resolution±0.1°C
Warranty3 Years
Dimensions (D x W x H)332 x 537 x 170 mm
Dimensions (D x W x H) Interior298 x 395 x 150 mm
Width (Metric)537 mm
Width (Metric) Interior395 mm
Height (Metric)170 mm
Height (Metric) Interior150 mm
Depth (Metric)332 mm
Depth (Metric) Interior298 mm
Weight (Metric)12 kg
Voltage230 V
Wattage1250 W