WS-200 Series Stuart™ Cabinet-Style Water Still
WS-200 Series Stuart™ Cabinet-Style Water Still
Actual product may vary
Fisherbrand
WS-200 Series Stuart™ Cabinet-Style Water Still
The WS-200 Series Stuart Cabinet-Style Water Still produces cold distilled water for immediate use.
Have Questions?
Catalog NumberOutput Flow RateDepth (Metric)
FKS114665384 L/hr.240 mm
Catalog number FKS11466538
Price (SGD)
17,931.00
EA
Add to cart
Output Flow Rate:
4 L/hr.
Depth (Metric):
240 mm
Price (SGD)
17,931.00
EA
Add to cart
  • Provides highest purity from virtually any raw water supply.
  • Reservoir water level sensor turns still off when reservoir is full.
  • Simple conversion to pretreated feed.
  • Safety features allow for unattended operation.
  • Remove organics, bacteria, and pyrogens from your water with WS-200 cabinet-style water stills.
  • Borosilicate glassware coupled with silica-sheathed heaters enclosed in a protective cabinet, delivers pyrogen-free distillate of the highest purity from virtually any raw water supply.
  • Stills are available in either single- or double-distilled water models—choose the double distilled model for higher purity levels.
  • The unique condenser design ensures that the distilled water droplets remain in contact with the cooling coil for an extended period for maximum energy transfer and immediate cold distilled water.
  • Safely and easily clean the unit without dismantling the glassware.
  • All-glass construction allows for rapid descaling using strong mineral acids.
  • A large bore stopcock with PTFE key is fitted for easy draining of chemicals after cleaning.
  • Safety features include flow sensor, overtemperature thermostat, acrylic screen, cabinet lid, and screw threads on all water connections.
  • A reservoir water level sensor automatically switches off the heaters and coolant water supply when the reservoir is filled with distilled water then automatically restarts when the distilled water level drops in the reservoir.
Specifications
Electrical Requirements220/240 V 50/60 Hz 3 kW
Frequency50/60 Hz
MaterialGlass
Phase1
Voltage240 V
Wattage3 kW
Inlet Water Temperature (Metric)25°C to 35°C
TypeWater Still
Output Flow Rate4 L/hr.
ModelA4000
Depth (English)9.4 in.
Depth (Metric)240 mm
Height (English)1.3 ft.
Height (Metric)410 mm
Width (English)1.8 ft.
Width (Metric)550 mm
Unit SizeEA