CultureGard™ Filter
Product Image
Fisherbrand
CultureGard™ Filter
Two CultureGard™ filters used in series provide the ultimate sterile barrier against the risk of culture contamination possible during media feeding and culture perfusion
Have Questions?
Catalog NumberPore Size
FKS114569980.2 μm
Catalog number FKS11456998
Price (SGD)
4,833.00
PK
Add to cart
Pore Size:
0.2 μm
Price (SGD)
4,833.00
PK
Add to cart
Two CultureGard™ filters used in series provide the ultimate sterile barrier against the risk of culture contamination possible during media feeding and culture perfusion. CultureGard™ autoclavable filters are made with 0.2μm MWCO DynaFibre™ membrane that permits the flow of culture medium while preventing the introduction of foreign microbials. These specialised filters have a large surface area for faster flow rates under lower pressures.
Components
HF membrane Mixed cellulose esterHF vent PolypropylenePotting material PolyurethaneHousing Clear polysulfoneEnd caps Pigmented polysulfone1/8inch HB adapters Polypropylene (4 MLL and 4 FLL)
  • Large surface area for fast flow rates under lower pressures
  • Minimal risk of membrane clogging or rupture
  • Minimum hold-up volume
  • PP vent fibre prevents air lock Hollow fibre sterilising grade filter for cell culture perfusion.
Specifications
MaterialPolysulfone
Pore Size0.2 μm
SterilitySterile
For Use With (Application)Used in series provide the ultimate sterile barrier against the risk of culture contamination possible during media feeding and culture perfusion
Quantity12Pack
TypeCultureGard Filter
Unit SizePK