Clear Bath
Clear Bath
Actual product may vary
Fisherbrand
Clear Bath
Keeps water baths, incubators, temperature controlled instruments, humidifiers, etc. clean and clear
Have Questions?
Catalog NumberVolume (Metric)
FKS11455839230 mL
Catalog number FKS11455839
Price (SGD)
-
Volume (Metric):
230 mL
  • Does not contain copper or other heavy metals
Contains State Restrictions
Specifications
FormLiquid
pH6.5 to 8.3
AppearancePale Yellow
Boiling Point200°F
Certifications/ComplianceUS OSHA, CFR 1910.1200, WHMIS
CompositionBenzalkonium Chloride, n-alkyl (50% C14, 40% C12, 10% C16) Dimethyl Benzyl Ammonium Chloride as acti
For Use With (Application)Used to keep any circulating, temperature-controlled instrument clean, instruments like circulating Water baths, incubators, temperature controlled instruments, humidifiers, automatic analyzer baths and Blood/gas analyzers
Volume (English)8 oz.
Volume (Metric)230 mL
Unit SizeEA