Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO
Spectra/Por&trade; 7 Membrane Tubing (Sulfur and Heavy Metal Free):<sup>1</sup> 1000 to 50,000 Dalton MWCO
Actual product may vary
Fisherbrand
Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)QuantityMWCO
FKS1144590929 mmEach2 kDa
FIS08-680-2E24 mmEach1 kDa
FKS1142587945 mmEach1 kDa
FIS08-680-3E24 mmEach2 kDa
FIS08-680-4E29 mmEach3.5 kDa
Catalog number FKS11445909
Price (SGD)
1,927.00
EA
Add to cart
Diameter (Metric):
29 mm
Quantity:
Each
MWCO:
2 kDa
Price (SGD)
1,927.00
EA
Add to cart
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Product TypeSpectra/Por 7 Membrane Tubing
MWCO2 kDa
MaterialRegenerated Cellulose
Diameter (Metric)29 mm
Diameter (English)1.14 in.
Length (English)16.4 ft.
Length (Metric)5 m
ColorTransparent
FormulationPackaged wet in 0.05% sodium azide
Temperature Range (English)39.2°F to 249.8°F
Temperature Range (Metric)4°C to 121°C
QuantityEach
For Use With (Application)Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Unit SizeEA